Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkab1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002632.1 | NP_001002632.1 | 2 - 3 | 258 | 87 | |
XM_005155628.4 | XP_005155685.1 | 2 - 3 | 325 | 87 | ||
GenBank mRNA | BC075947.1 | - | 2 - 3 | 258 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |