Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003869.1 | NP_001003869.1 | 5 - 6 | 1104 | 68 | |
GenBank mRNA | AM422111.2 | - | 2 - 3 | 265 | 68 | |
AY648791.1 | - | 5 - 6 | 1104 | 68 | ||
BC093104.1 | - | 5 - 6 | 1051 | 68 | ||
BC115114.1 | - | 5 - 6 | 1055 | 68 | ||
BC121728.1 | - | 5 - 6 | 1023 | 68 | ||
BC129281.1 | - | 5 - 6 | 1053 | 68 | ||
BC134116.1 | - | 5 - 6 | 1052 | 68 | ||
BC164482.1 | - | 4 - 5 | 940 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |