Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: krcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012366.1 | NP_001012366.1 | 5 - 6 | 682 | 62 | |
XM_005172254.4 | XP_005172311.1 | 5 - 6 | 758 | 62 | ||
GenBank mRNA | AY835383.1 | - | 5 - 6 | 682 | 62 | |
BC108043.1 | - | 5 - 6 | 746 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |