Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exosc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017585.1 | NP_001017585.1 | 6 - 7 | 648 | 89 | |
GenBank mRNA | BC092997.1 | - | 6 - 7 | 648 | 89 | |
BC152123.1 | - | 6 - 7 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |