Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020572.2 | NP_001018408.1 | 5 - 6 | 567 | 69 | |
XM_021470172.1 | XP_021325847.1 | 5 - 6 | 590 | 69 | ||
GenBank mRNA | BC095198.1 | - | 5 - 6 | 567 | 69 | |
BC165539.1 | - | 5 - 6 | 489 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |