Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020729.1 | NP_001018565.1 | 1 - 2 | 202 | 79 | |
XM_017358228.2 | XP_017213717.1 | 1 - 2 | 222 | 79 | ||
GenBank mRNA | BC072723.1 | - | 1 - 2 | 174 | 79 | |
BC095064.1 | - | 1 - 2 | 202 | 79 | ||
BC164759.1 | - | 1 - 2 | 148 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |