Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042735.1 | NP_001036200.1 | 6 - 7 | 1795 | 80 | |
GenBank mRNA | BC162801.1 | - | 6 - 7 | 1895 | 80 | |
BC162802.1 | - | 6 - 7 | 1895 | 80 | ||
DQ641948.1 | - | 6 - 7 | 1795 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |