Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: senp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080197.2 | NP_001073666.2 | 5 - 6 | 1262 | 80 | |
GenBank mRNA | BC155797.1 | - | 4 - 5 | 415 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |