Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mxra8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080582.1 | NP_001074051.1 | 1 - 2 | 188 | 61 | |
GenBank mRNA | BC129223.1 | - | 1 - 2 | 188 | 61 | |
CU638765.1 | - | 1 - 2 | 188 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |