Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082803.1 | NP_001076272.1 | 10 - 11 | 1395 | 69 | |
GenBank mRNA | BC124310.1 | - | 10 - 11 | 1420 | 69 | |
BC133897.1 | - | 10 - 11 | 1395 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |