Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083572.2 | NP_001077041.1 | 2 - 3 | 271 | 115 | |
GenBank mRNA | BC134222.1 | - | 2 - 3 | 272 | 115 | |
FJ032031.1 | - | 2 - 3 | 248 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |