Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfkmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089417.2 | NP_001082886.2 | 7 - 8 | 824 | 130 | |
NM_001365336.1 | NP_001352265.1 | 10 - 11 | 1338 | 130 | ||
GenBank mRNA | BC139532.1 | - | 7 - 8 | 811 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |