Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftr02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003197921.5 | XP_003197969.1 | 1 - 2 | 691 | 87 | |
GenBank mRNA | BC115273.1 | - | 1 - 2 | 630 | 87 | |
CK027015.1 | - | 1 - 2 | 625 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |