Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: es1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131039.1 | NP_571114.1 | 1 - 2 | 182 | 93 | |
GenBank mRNA | BC122433.1 | - | 1 - 2 | 163 | 93 | |
U10403.1 | - | 1 - 2 | 182 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |