Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dachb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152956.2 | NP_694488.2 | 2 - 3 | 685 | 97 | |
XM_009305776.3 | XP_009304051.1 | 2 - 3 | 716 | 97 | ||
GenBank mRNA | AF427109.1 | - | 2 - 3 | 659 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |