Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serpinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182863.1 | NP_878283.1 | 7 - 8 | 1284 | 88 | |
GenBank mRNA | AF515272.1 | - | 7 - 8 | 1284 | 88 | |
BC081642.1 | - | 7 - 8 | 1246 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |