Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ahcy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199218.1 | NP_954688.1 | 9 - 10 | 1285 | 117 | |
GenBank mRNA | AY398307.1 | - | 9 - 10 | 1285 | 117 | |
BC044200.1 | - | 9 - 10 | 1253 | 117 | ||
BC164526.1 | - | 9 - 10 | 1211 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |