Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200736.1 | NP_957030.1 | 4 - 5 | 424 | 64 | |
GenBank mRNA | BC059520.1 | - | 4 - 5 | 424 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |