Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bysl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201106.1 | NP_957400.1 | 5 - 6 | 881 | 99 | |
GenBank mRNA | AY648821.1 | - | 5 - 6 | 903 | 99 | |
BC053134.1 | - | 5 - 6 | 881 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |