Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alcamb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212634.3 | NP_997799.3 | 5 - 6 | 685 | 70 | |
XM_005157504.4 | XP_005157561.1 | 5 - 6 | 696 | 70 | ||
GenBank mRNA | AY050561.1 | - | 5 - 6 | 641 | 70 | |
BC163458.1 | - | 5 - 6 | 591 | 70 | ||
BC163486.1 | - | 5 - 6 | 591 | 70 | ||
CU458904.1 | - | 5 - 6 | 555 | 70 | ||
CU458936.1 | - | 5 - 6 | 607 | 70 | ||
DQ279080.1 | - | 5 - 6 | 648 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |