Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007400.2 | NP_001007401.2 | 8 - 9 | 819 | 161 | |
XM_017354073.2 | XP_017209562.1 | 7 - 8 | 900 | 161 | ||
GenBank mRNA | BC085438.1 | - | 8 - 9 | 821 | 161 | |
BC165210.1 | - | 7 - 8 | 684 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |