Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: morn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017559.1 | NP_001017559.1 | 4 - 5 | 425 | 56 | |
GenBank mRNA | BC093438.1 | - | 4 - 5 | 425 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |