Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stxbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025182.1 | NP_001020353.1 | 3 - 4 | 188 | 76 | |
XM_005161113.4 | XP_005161170.1 | 3 - 4 | 263 | 76 | ||
GenBank mRNA | BC097221.1 | - | 3 - 4 | 188 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |