Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfsf10l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042713.1 | NP_001036178.1 | 2 - 3 | 286 | 92 | |
GenBank mRNA | BC162834.1 | - | 2 - 3 | 368 | 92 | |
BC162841.1 | - | 2 - 3 | 368 | 92 | ||
DQ812113.1 | - | 2 - 3 | 286 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |