Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem54b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044716.2 | NP_001038181.2 | 4 - 5 | 566 | 94 | |
GenBank mRNA | BC134098.1 | - | 4 - 5 | 566 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |