Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082807.1 | NP_001076276.1 | 4 - 5 | 832 | 97 | |
GenBank mRNA | BC134198.1 | - | 4 - 5 | 832 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |