Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104649.2 | NP_001098119.2 | 13 - 14 | 1312 | 138 | |
XM_021471707.1 | XP_021327382.1 | 12 - 13 | 1285 | 138 | ||
XM_021471709.1 | XP_021327384.1 | 11 - 12 | 1075 | 138 | ||
XM_021471716.1 | XP_021327391.1 | 13 - 14 | 1374 | 138 | ||
GenBank mRNA | BC163912.1 | - | 13 - 14 | 1334 | 138 | |
EF653429.1 | - | 13 - 14 | 1394 | 138 | ||
EU124003.1 | - | 13 - 14 | 1314 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |