Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp17a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105670.1 | NP_001099140.1 | 1 - 2 | 187 | 84 | |
GenBank mRNA | EF624003.1 | - | 1 - 2 | 187 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |