Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244600.1 | NP_001231529.1 | 7 - 8 | 1390 | 77 | |
NM_001244601.1 | NP_001231530.1 | 7 - 8 | 1264 | 77 | ||
NM_131453.3 | NP_571528.2 | 7 - 8 | 1025 | 77 | ||
XM_005166786.4 | XP_005166843.1 | 6 - 7 | 1302 | 77 | ||
XM_017357272.2 | XP_017212761.1 | 6 - 7 | 1141 | 77 | ||
XM_021478178.1 | XP_021333853.1 | 7 - 8 | 1379 | 77 | ||
GenBank mRNA | BC065965.1 | - | 7 - 8 | 1025 | 77 | |
BC095018.1 | - | 5 - 6 | 664 | 77 | ||
U17602.1 | - | 6 - 7 | 871 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |