Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dixdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182884.1 | NP_878304.1 | 10 - 11 | 1270 | 96 | |
GenBank mRNA | AF502143.1 | - | 10 - 11 | 1270 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |