Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198916.2 | NP_944598.2 | 1 - 2 | 671 | 116 | |
GenBank mRNA | AY331582.1 | - | 1 - 2 | 593 | 116 | |
BC066726.1 | - | 1 - 2 | 671 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |