Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201052.1 | NP_957346.1 | 4 - 5 | 582 | 120 | |
GenBank mRNA | AY391434.1 | - | 4 - 5 | 563 | 120 | |
BC044504.1 | - | 4 - 5 | 582 | 120 | ||
BC062859.1 | - | 4 - 5 | 565 | 120 | ||
BC164084.1 | - | 3 - 4 | 531 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |