Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppm1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201488.1 | NP_958896.1 | 10 - 11 | 1693 | 135 | |
XM_005169512.4 | XP_005169569.1 | 9 - 10 | 1861 | 261 | ||
GenBank mRNA | BC052132.1 | - | 10 - 11 | 1693 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |