Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam49bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207078.1 | NP_996961.1 | 8 - 9 | 786 | 69 | |
GenBank mRNA | BC066542.1 | - | 8 - 9 | 786 | 69 | |
BC152197.1 | - | 8 - 9 | 850 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |