Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: per1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212439.2 | NP_997604.2 | 4 - 5 | 1363 | 117 | |
GenBank mRNA | AY597250.1 | - | 4 - 5 | 1363 | 117 | |
BC163718.1 | - | 3 - 4 | 908 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |