Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pwp2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213047.1 | NP_998212.1 | 15 - 16 | 2086 | 124 | |
GenBank mRNA | BC047817.1 | - | 15 - 16 | 2086 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |