Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bambib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002189.1 | NP_001002189.1 | 2 - 3 | 646 | 71 | |
GenBank mRNA | BC072715.1 | - | 2 - 3 | 646 | 71 | |
BC165294.1 | - | 2 - 3 | 395 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |