Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002405.1 | NP_001002405.1 | 15 - 16 | 1159 | 64 | |
GenBank mRNA | AY900006.1 | - | 10 - 11 | 911 | 64 | |
BC076177.1 | - | 15 - 16 | 1159 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |