Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epb41l3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003987.1 | NP_001003987.1 | 9 - 10 | 1087 | 106 | |
XM_021470142.1 | XP_021325817.1 | 10 - 11 | 1199 | 106 | ||
XM_021470143.1 | XP_021325818.1 | 9 - 10 | 1153 | 106 | ||
XM_021470144.1 | XP_021325819.1 | 10 - 11 | 1198 | 106 | ||
XM_021470145.1 | XP_021325820.1 | 10 - 11 | 1191 | 106 | ||
XM_021470146.1 | XP_021325821.1 | 10 - 11 | 1371 | 106 | ||
XM_021470147.1 | XP_021325822.1 | 10 - 11 | 1230 | 106 | ||
XM_021470148.1 | XP_021325823.1 | 11 - 12 | 1311 | 106 | ||
XM_021470149.1 | XP_021325824.1 | 10 - 11 | 1148 | 106 | ||
XM_021470150.1 | XP_021325825.1 | 10 - 11 | 1196 | 106 | ||
XM_021470151.1 | XP_021325826.1 | 10 - 11 | 1195 | 106 | ||
XM_021470152.1 | XP_021325827.1 | 10 - 11 | 1197 | 106 | ||
XM_021470153.1 | XP_021325828.1 | 9 - 10 | 1146 | 106 | ||
XM_021470154.1 | XP_021325829.1 | 9 - 10 | 1093 | 106 | ||
GenBank mRNA | BC080246.1 | - | 9 - 10 | 1087 | 106 | |
BC158197.1 | - | 9 - 10 | 1093 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |