Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcnip1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008632.2 | NP_001008632.2 | 6 - 7 | 675 | 73 | |
NM_001328504.1 | NP_001315433.1 | 6 - 7 | 819 | 73 | ||
XM_021479166.1 | XP_021334841.1 | 6 - 7 | 1138 | 73 | ||
GenBank mRNA | BC086698.1 | - | 6 - 7 | 675 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |