Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030123.1 | NP_001025294.1 | 28 - 29 | 4039 | 75 | |
XM_009294751.3 | XP_009293026.1 | 28 - 29 | 4060 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |