Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr6a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033720.1 | NP_001028892.1 | 6 - 7 | 809 | 59 | |
GenBank mRNA | BC097237.1 | - | 6 - 7 | 809 | 59 | |
DQ017610.1 | - | 4 - 5 | 622 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |