Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pi4kaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034972.1 | NP_001030144.1 | 25 - 26 | 2889 | 104 | |
XM_005172596.4 | XP_005172653.1 | 26 - 27 | 3117 | 104 | ||
XM_009305081.3 | XP_009303356.1 | 26 - 27 | 3118 | 104 | ||
GenBank mRNA | AY929292.1 | - | 25 - 26 | 2889 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |