Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slco3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045188.2 | NP_001038653.1 | 6 - 7 | 1318 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |