Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thbs3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045214.1 | NP_001038679.1 | 18 - 19 | 2176 | 64 | |
XM_005170062.4 | XP_005170119.1 | 18 - 19 | 2587 | 64 | ||
GenBank mRNA | BC163250.1 | - | 17 - 18 | 2134 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |