Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nfkbie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080089.1 | NP_001073558.1 | 5 - 6 | 1215 | 84 | |
GenBank mRNA | BC128831.1 | - | 5 - 6 | 1215 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |