Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxo1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082857.1 | NP_001076326.1 | 2 - 3 | 755 | 75 | |
GenBank mRNA | BC163020.1 | - | 2 - 3 | 787 | 75 | |
EF421425.1 | - | 2 - 3 | 755 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |