Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loxl5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102574.1 | NP_001096044.1 | 4 - 5 | 1119 | 81 | |
XM_017357369.2 | XP_017212858.1 | 4 - 5 | 1271 | 81 | ||
XM_017357372.2 | XP_017212861.1 | 5 - 6 | 1736 | 81 | ||
GenBank mRNA | EF030486.1 | - | 4 - 5 | 1119 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |