Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: htatip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131681.2 | NP_571756.1 | 4 - 5 | 650 | 79 | |
NR_131978.1 | 3 - 4 | 1869 | 79 | |||
GenBank mRNA | AF329850.1 | - | 4 - 5 | 619 | 79 | |
BC081572.1 | - | 3 - 4 | 1807 | 79 | ||
BC097086.1 | - | 4 - 5 | 650 | 79 | ||
EH476941.1 | - | 2 - 3 | 557 | 79 | ||
EH550887.1 | - | 4 - 5 | 687 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |