Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: caspb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152884.2 | NP_690840.2 | 6 - 7 | 1029 | 77 | |
GenBank mRNA | AF327410.1 | - | 5 - 6 | 992 | 77 | |
BC095000.1 | - | 6 - 7 | 1029 | 77 | ||
BC164498.1 | - | 5 - 6 | 1026 | 77 | ||
DQ022755.1 | - | 5 - 6 | 992 | 77 | ||
MG958005.1 | - | 5 - 6 | 992 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |